Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bax Antibody (SPM336), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB279135 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB279135 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB279135 Supplier Novus Biologicals Supplier No. NBP234763MFV450
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Bax Monoclonal antibody specifically detects Bax in Human, Monkey, Mouse (Negative) samples. It is validated for Flow Cytometry

Specifications

Antigen Bax
Applications Flow Cytometry
Classification Monoclonal
Clone SPM336
Conjugate mFluor Violet 450 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias apoptosis regulator BAX, BCL2-associated X protein, Bcl2-L-4, BCL2L4bcl2-L-4, Bcl-2-like protein 4
Host Species Mouse
Immunogen A synthetic peptide, aa 3-16 (Cys-GSGEQPRGGGPTSS) of human bax protein. (Uniprot: Q07812)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Core ESC Like Genes, Stem Cell Markers, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 581
Target Species Human, Monkey, Mouse (Negative)
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.