Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bax inhibitor 1 Antibody (20F430), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP224919AF750
Description
Bax inhibitor 1 Monoclonal antibody specifically detects Bax inhibitor 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
Bax inhibitor 1 | |
Monoclonal | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
20F430 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
BAXI1, BI1, BI-1Transmembrane BAX inhibitor motif-containing protein 6, TEGTbax inhibitor 1, testis enhanced gene transcript, Testis-enhanced gene transcript protein, transmembrane BAX inhibitor motif containing 6 | |
A synthetic peptide made to the N-terminal portion of the human Bax inhibitor 1 protein (between amino acids 1-40) UniProt P55061. | |
0.1 mL | |
Apoptosis | |
7009 | |
Store at 4C in the dark. | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction