Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bax inhibitor 1 Antibody (20F430), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP224919G
Description
Bax inhibitor 1 Monoclonal specifically detects Bax inhibitor 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Bax inhibitor 1 | |
Monoclonal | |
DyLight 488 | |
P55061 | |
TMBIM6 | |
A mixture of three synthetic peptides corresponding to the N-terminal, internal and C-terminal regions of human B1. Epitope mapping indicates that the antibody recognizes an epitope in the N-terminal region of Bi-1. | |
0.1 mL | |
Apoptosis | |
7009 | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
20F430 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
BAXI1, BI1, BI-1Transmembrane BAX inhibitor motif-containing protein 6, TEGTbax inhibitor 1, testis enhanced gene transcript, Testis-enhanced gene transcript protein, transmembrane BAX inhibitor motif containing 6 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
This antibody is reactive to Human, Mouse, Rat. | |
Store at 4C in the dark. | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction