Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bax inhibitor 1 Antibody (20F430), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 2 publications
$192.47 - $517.59
Specifications
| Antigen | Bax inhibitor 1 |
|---|---|
| Clone | 20F430 |
| Concentration | 1.0 mg/ml |
| Dilution | Western Blot reported in scientific literature (PMID 20359348), Immunohistochemistry, Immunohistochemistry-Paraffin |
| Applications | Immunohistochemistry (Paraffin) |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP224919SS
![]() |
Novus Biologicals
NBP224919SS |
0.025 mg |
Each for $192.47
|
|
|||||
NBP224919
![]() |
Novus Biologicals
NBP224919 |
0.1 mg |
Each for $517.59
|
|
|||||
Description
Bax inhibitor 1 Monoclonal specifically detects Bax inhibitor 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Bax inhibitor 1 | |
| 1.0 mg/ml | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Apoptosis | |
| P55061 | |
| 7009 | |
| A synthetic peptide made to the N-terminal portion of the human Bax inhibitor 1 protein (between amino acids 1-40) UniProt P55061. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 20F430 | |
| Western Blot reported in scientific literature (PMID 20359348), Immunohistochemistry, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| BAXI1, BI1, BI-1Transmembrane BAX inhibitor motif-containing protein 6, TEGTbax inhibitor 1, testis enhanced gene transcript, Testis-enhanced gene transcript protein, transmembrane BAX inhibitor motif containing 6 | |
| TMBIM6 | |
| IgG2b κ | |
| Protein G purified | |
| This antibody is reactive to Human, Mouse, Rat. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title