Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Bax Monoclonal Antibody (SP47)
GREENER_CHOICE

Catalog No. PIMA516322
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA516322 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA516322 Supplier Invitrogen™ Supplier No. MA516322
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10-20 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 30 min at room temperature. A suggested positive control is normal breast tissue.

BAX is a members of the Bcl-2 Family and plays an important role in regulation of apoptosis. Whereas Bcl-2 is commonly regarded as an anti-apoptotic protein, BAX is considered to have a pro-apoptotic function. Regulation of apoptosis is supposed to involve both homo- and heterodimerization of different isoforms of BAX and Bcl-2. The Bax gene encodes different isoforms including Bax alpha (21 kDa) and Bax beta (24 kDa), whereas both isoforms contain the BH1, BH2 and BH3 domains, Bax beta has a unique carboxyl terminus and does not contain a hydrophobic transmembrane domain. Bcl-2 is also expressed in different Isoforms. Bcl-2 beta differs in the 3' UTR and coding region compared to variant alpha. Bcl-2 beta is shorter (22 kDa) and has a distinct C-terminus compared to Bcl-2 alpha (26 kDa). BAX is a member of the BCL-2 family of proteins, which function as regulators of apoptosis. Overexpression of BAX functions to promote cell death. BAX can form homodimers and is also able to heterodimerize with other BCL-2 related proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Bax
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SP47
Conjugate Unconjugated
Formulation PBS with 1% BSA and 0.1% sodium azide; pH 7.6
Gene BAX
Gene Accession No. Q07812
Gene Alias apoptosis regulator BAX; Apoptosis regulator Bcl-X; Bax; Bax zeta; BAX-ALPHA; Bax-alpha protein; Baxdelta2G9; Baxdelta2G9omega; Baxdelta2omega; BCL2 associated X protein; BCL2 associated X, apoptosis regulator; BCL2 like 1; BCL2-associated protein; Bcl-2-associated protein Bax; Bcl2-associated protein Bax; Bcl-2-associated X protein; BCL2-associated X protein; BCL2-associated X protein omega; BCL2L; BCL2L1; Bcl2-L-1; BCL2L4; Bcl2-L-4; BCL2-like 1; bcl-2-like protein 1; Bcl-2-like protein 4; BCLX; Bcl-X; BCL-XL/S; PPP1R52; protein phosphatase 1, regulatory subunit 52
Gene Symbols BAX
Host Species Rabbit
Immunogen Synthetic peptide corresponding to internal region of human BAX protein.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 581
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.