Learn More
BAX Monoclonal Antibody (ZY455), Abbomax
Mouse Monoclonal Antibody
Supplier: Abbomax Inc 604090

Description
Positive control: Human lymph node Cellular location: Cytoplasm and Mitochondria membrane.
BAX Monoclonal antibody specifically detects BAX in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
BAX | |
Monoclonal | |
0.5 mg/mL | |
PBS with proprietary stabilizer and 0.01% sodium azide | |
Q07812, Q07813, Q63690 | |
Mouse | |
Protein A | |
RUO | |
12028, 24887, 581 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
ZY455 | |
Unconjugated | |
BAX | |
apoptosis regulator BAX; Apoptosis regulator Bcl-X; Bax; Bax zeta; BAX-ALPHA; Bax-alpha protein; Baxdelta2G9; Baxdelta2G9omega; Baxdelta2omega; BCL2 associated X protein; BCL2 associated X, apoptosis regulator; BCL2 like 1; BCL2-associated protein; Bcl-2-associated protein Bax; Bcl2-associated protein Bax; Bcl-2-associated X protein; BCL2-associated X protein; BCL2-associated X protein omega; BCL2L; BCL2L1; Bcl2-L-1; BCL2L4; Bcl2-L-4; BCL2-like 1; bcl-2-like protein 1; Bcl-2-like protein 4; BCLX; Bcl-X; BCL-XL/S; PPP1R52; protein phosphatase 1, regulatory subunit 52 | |
A recombinant protein of full length human BAX | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.