Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Bay 11-7821
SDP

Catalog No. 174410 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-441-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 17-441-0 Supplier Tocris Bioscience™ Supplier No. 1744/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

E2 ubiquitin (Ub) conjugating enzymes inhibitor

E2 ubiquitin (Ub) conjugating enzyme inhibitor. Inhibits the conjugation of Ub to a range of E2 enzymes. Inhibits E2 enzyme mediated IκBα phosphorylation and indirectly decreases NF-κb activity. Also reversibly activates MAPK signaling. Induces apoptosis of leukemic T cell and B-cell lymphoma cell lines. Also inhibits activation of NLRP3 inflammasome.

Chemical Identifiers

CAS 19542-67-7
Molecular Formula C10H9NO2S

Specifications

Quantity 10 mg
Formula Weight 207.25
Purity >99%
Molecular Weight (g/mol) 207.25
Chemical Name or Material Bay 11-7821
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.