Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ BAY 299
GREENER_CHOICE

Catalog No. 597025
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
59-702-5 25 mg
59-705 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 59-702-5 Supplier Tocris Bioscience™ Supplier No. 5970/25
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective BRD1 and TAF1 inhibitor

BAY 299 is a potent and selective BRD1 and TAF1 inhibitor (IC50 values are 6-67 and 8-13 nM, respectively). Displays selectivity over other bromodomains (>30-fold over other members of the BRPF family; BRD9 and ATAD2; >300-fold over BRD4). Displays BRD1 and TAF1 inhibition in a NanoBRET cell assay. Inhibits binding of BRD1 and TAF1 to histone H4 (IC50 values are 575 nM and 0.9 μM, respectively) and histone H3.3 (IC50 values are 825 nM and 1.4 μM, respectively).

Specifications

Chemical Name or Material 6-(3-Hydroxypropyl)-2-(1,3,6-trimethyl-2-oxo-2,3-dihydro-1H-benzimidazol-5-yl)-1H-benzo[de]isoquinoline-1,3(2H)-dione
CAS 2080306-23-4
Assay Percent Range >98%
Quantity 25 mg
Molecular Formula C25H23N3O4
Formula Weight Observed MW: 429.47
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.