Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ BAY 41-8543, >98%
GREENER_CHOICESDP

Catalog No. 570810 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
57-081-0 10 mg
57-085-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 57-081-0 Supplier Tocris Bioscience™ Supplier No. 5708/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Guanylyl cyclase (sGC) stimulator

Guanylyl cyclase (sGC) stimulator. Increases activity of sGC by up to 92-fold at concentrations up to 100 μM. Induces relaxation of aorta, saphenous arteries, coronary arteries and veins in vitro. Inhibits collagen-induced platelet aggregation. Promotes lipid uptake into brown adipose tissue and increases energy expenditure in mice. Induces weight loss in obese mice. Antihypertensive.

Chemical Identifiers

CAS 256498-66-5
Molecular Formula C21H21FN8O

Specifications

Quantity 10 mg
Solubility Soluble to 10mM in DMSO with gentle warming
Formula Weight 420.44
Purity >98%
Molecular Weight (g/mol) 420.452
Chemical Name or Material 2-[1-[(2-Fluorophenyl)methyl]-1H-pyrazolo[3,4-b]pyridin-3-yl]-5-(4-morpholinyl)-4,6-pyrimidinediamin
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.