Learn More
Description
To request the negative control for BAY 598, please fill out the BAY 369 request form on the SGC website.
Specifications
Specifications
| Chemical Name or Material | N-[(4 S)-1-[(Cyanoamino)[[3-(difluoromethoxy)phenyl]imino]methyl]-3-(3,4-dichlorophenyl)-4,5-dihydro-1 H-pyrazol-4-yl]-N-ethyl-2-hydroxyacetamide |
| CAS | 1906919-67-2 |
| Quantity | 5 mg |
| Target | Additional Lysine Methyltransferase Compounds |
| Molecular Formula | C22H20Cl2F2N6O3 |
| Formula Weight | 525.34 |
| Purity | 98% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
