Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ BAY 6096
SDP

Catalog No. 800650 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
80-065-0 50 mg
80-061-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 80-065-0 Supplier Tocris Bioscience™ Supplier No. 8006/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Potent and selective adrenergic α2B antagonist

BAY 6096 is a potent (IC50 = 14 nM), and selective adrenergic α2B receptor antagonist that displays 394, 725, >845-fold selectivity over α1A, α2A, and α2C, respectively. BAY 6096 reduces α2B receptor agonist-induced vasoconstriction and has a high clearance and low mean residence times in rats. BAY 6096 is highly water-soluble.

Specifications

Chemical Name or Material 1-[2-[[[3-(3,5-Dimethyl-4-isoxazolyl)imidazo[1,2-a]pyridin-7-yl]carbonyl]amino]ethyl]-4-(methylamino)pyridinium chloride
CAS 2313483-41-7
Quantity 50 mg
Target Adrenergic alpha-2 Receptor Antagonists
Molecular Formula C21H23ClN6O2
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.