Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ BAY 678
GREENER_CHOICE

Catalog No. 570625
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
57-062-5 25 mg
57-065 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 57-062-5 Supplier Tocris Bioscience™ Supplier No. 5706/25
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective cell-permeable human neutrophil elastase (HNE) inhibitor

BAY 678 is a potent and selective cell-permeable human neutrophil elastase (HNE) inhibitor (IC50 = 20 nM). Exhibits around 2000-fold selectivity for NHE against a panel of 21 other serine proteases. Effective in an acute lung injury model in vivo. Orally bioavailable. Inactive S-enantiomer (Cat.No.6389) also available.

Specifications

Chemical Name or Material 5-[(4R)-5-Acetyl-1,2,3,4-tetrahydro-6-methyl-2-oxo-1-[3-(trifluoromethyl)phenyl]-4-pyrimidinyl]-2-pyridinecarbonitrile
CAS 675103-36-3
Assay Percent Range >98%
Quantity 25 mg
Molecular Formula C20H15F3N4O2
Formula Weight Observed MW: 400.35
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.