Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ BAY 805
SDP

Catalog No. 798450 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
79-845-0 50 mg
79-841-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 79-845-0 Supplier Tocris Bioscience™ Supplier No. 7984/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective inhibitor of USP21; also NF-κB activator

BAY 805 is a potent and selective inhibitor of USP21 (IC50 = 6 nM, Kd = 2.2 nM). BAY 805 induces NF-κB activation (EC50= 17 nM in a cell-based reporter assay).

Specifications

Chemical Name or Material N-[(1R)-1-[[[5-[(4-Cyanophenyl)methyl]-1,3,4-thiadiazol-2-yl]amino]carbonyl]-3-methylbutyl]-1-(trifluoromethyl)cyclohexanecarboxamide
CAS 2925481-88-3
Quantity 50 mg
Target Deubiquitinating Enzyme Inhibitors
Molecular Formula C24H28F3N5O2S
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.