Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ BAY 876
SDP

Catalog No. 61995 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
61-995 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 61-995 Supplier Tocris Bioscience™ Supplier No. 6199/5
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective GLUT1 inhibitor

BAY 876 is a potent and selective GLUT1 inhibitor (IC50 = 2 nM) that displays selectivity for GLUT1 over GLUT2/3/4 (IC50 values are 10.8, 1.67 and 0.29 μM, respectively). BAY 876 induces cell death in hypoxic conditions in vitro and inhibits glucose uptake by Hela-MaTu cells. In ovarian cancer, BAY 876 reduces glycolysis rates and ATP production and inhibits proliferation in vitro and in vivo. This compound is cell permeable and orally bioavailable.

Specifications

Chemical Name or Material N4-[1-[(4-Cyanophenyl)methyl]-5-methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl]-7-fluoro-2,4-quinolinedicarboxamide
CAS 1799753-84-6
Quantity 5 mg
Molecular Formula C24H16F4N6O2
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.