Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ BAY-u 3405
GREENER_CHOICE

Catalog No. 273210
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
27-321-0 10 mg
27-325-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 27-321-0 Supplier Tocris Bioscience™ Supplier No. 2732/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Dual TP/DP2 (CRTH2) receptor antagonist

Potent dual antagonist of TP/DP2 (CRTH2) prostanoid receptors (Ki values are 4.3, 4.5 and > 10000 nM for hDP2, hTP and hDP1 receptors respectively). Suppresses PGD2-induced migration of human eosinophils (IC50 = 170 nM).

Chemical Identifiers

CAS 116649-85-5
Molecular Formula C21H21FN2O4S

Specifications

Quantity 10 mg
Formula Weight 416.47
Purity >99%
Molecular Weight (g/mol) 416.467
Chemical Name or Material BAY-u 3405
Recommended Storage Desiccate at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.