Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ BAY-u 3405
GREENER_CHOICESDP

Catalog No. 273250 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
27-325-0 50 mg
27-321-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 27-325-0 Supplier R&D Systems™ Supplier No. 2732/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Dual TP/DP2 (CRTH2) receptor antagonist

Potent dual antagonist of TP/DP2 (CRTH2) prostanoid receptors (Ki values are 4.3, 4.5 and > 10000 nM for hDP2, hTP and hDP1 receptors respectively). Suppresses PGD2-induced migration of human eosinophils (IC50 = 170 nM).

Chemical Identifiers

CAS 116649-85-5
Molecular Formula C21H21FN2O4S

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 416.47
Purity >99%
Molecular Weight (g/mol) 416.467
Chemical Name or Material BAY-u 3405
Recommended Storage Desiccate at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.