Learn More
Ascensia Diabetes Care ACETEST™ Reagent Tablets
Catalog No. am2381
Click to view available options
Quantity:
100 Tablets
This item is not returnable. View return policy
Catalog No. AM-2381
Supplier: Ascensia Diabetes Care 2381
This item is not returnable. View return policy
Contain several ingredients for semi-quantitative measurement of ketones in urine
  • Also test serum plasma and whole blood
  • Bottle of 100 tablets
Specifications
Clia Complexity Waived
Type Acetest™ Reagent tTablet
For Use With (Application) Ideal for semi-quantitative measurement of Ketones in Urine and also test Serum Plasma and whole Blood
Quantity 100 Tablets
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.