Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BAZ1A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597272
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597272 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597272 Supplier Invitrogen™ Supplier No. PA597272
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VFVELLSPRR KRRGRKSANN TPENSPNFPN FRVIATKSSE QSRSVNIASK LSLQESESKR RCRKRQSPEP SPVTLGRRSS GRQGGVHELS AFEQLVVELV RHDDSWPFLK LVSKIQVPDY YDIIKKPIAL NIIREKVNKC EYKLASEFID DIELMFSNCF EYNPRNTSEA KAGTRLQAFF HIQAQKLGLH VTPSNVDQVS TPPAAKKSRI; Positive Samples: HeLa, Mouse lung, Mouse spleen, Rat lung, Rat skeletal muscle; Cellular Location: Nucleus.

BAZ1A may play a role in transcriptional regulation. May be involved in the formation or maintenance of heterochromatin playing a critical role in developmental control.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BAZ1A
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene BAZ1A
Gene Accession No. O88379, Q9NRL2
Gene Alias ACF1; ATP-dependent chromatin remodeling protein; ATP-dependent chromatin-remodeling protein; ATP-utilizing chromatin assembly and remodeling factor 1; B930060C03; Baz1a; BC065123; bromodomain adjacent to zinc finger domain 1A; bromodomain adjacent to zinc finger domain protein 1A; bromodomain adjacent to zinc finger domain, 1A; cbp146; CHRAC subunit ACF1; gene trap locus 5; Gtl5; hACF1; HSPC317; hWALp1; WALp1; WCRF180; Williams syndrome transcription factor-related chromatin-remodeling factor 180
Gene Symbols BAZ1A
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1347-1556 of human BAZ1A (NP_0384762).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11177, 217578, 314126
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.