Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BAZ1B Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA566889

Catalog No. PIPA566889


Only null left
Add to Cart

Description

Description

Immunogen sequence: RIRKHKAAAEK AFQEGIAKAK LVMRRTPIGT DRNHNRYWLF SDEVPGLFIE KGWVHDSIDY RFNHHCKDHT VSGDE Highest antigen sequence identity to the following orthologs - mouse 86%, rat 86%.

BAZ1B is a member of the bromodomain protein family. The bromodomain is a structural motif characteristic of proteins involved in chromatin-dependent regulation of transcription. This gene is deleted in Williams-Beuren syndrome, a developmental disorder caused by deletion of multiple genes at 7q11.23.This gene encodes a member of the bromodomain protein family. The bromodomain is a structural motif characteristic of proteins involved in chromatin-dependent regulation of transcription. This gene is deleted in Williams-Beuren syndrome, a developmental disorder caused by deletion of multiple genes at 7q11.23.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

BAZ1B
Polyclonal
Unconjugated
BAZ1B
BAZ1B; bromodomain adjacent to zinc finger domain 1B; bromodomain adjacent to zinc finger domain protein 1B; bromodomain adjacent to zinc finger domain, 1B; bromodomain adjacent to zinc finger domain, 1B; Williams-Beuren syndrome chromosome region 9 homolog; C87820; hWALp2; transcription factor WSTF; tyrosine-protein kinase BAZ1B; tyrosine-protein kinase BAZ1B; LOW QUALITY PROTEIN: tyrosine-protein kinase BAZ1B; Tyrosine-protein kinase BAZ1B-like protein; WBSC10; WBSCR10; WBSCR9; williams syndrome transcription factor; Williams syndrome transcription factor homolog; williams-Beuren syndrome chromosomal region 10 protein; Williams-Beuren syndrome chromosomal region 9 protein; williams-Beuren syndrome chromosomal region 9 protein homolog; Williams-Beuren syndrome chromosome region 9 homolog; Wstf
Rabbit
Antigen affinity chromatography
RUO
9031
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry
0.3 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9UIG0
BAZ1B
Recombinant Human BAZ1B. Recombinant protein control fragment (Product #RP-107536).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.