Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BAZ1B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5121245
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121245 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5121245 Supplier Invitrogen™ Supplier No. PA5121245
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: SW480, BT-474, SKOV3, HeLa, HepG2, Mouse thymus. The target is usually found in the following locations: Nucleus.

BAZ1B is a member of the bromodomain protein family. The bromodomain is a structural motif characteristic of proteins involved in chromatin-dependent regulation of transcription. This gene is deleted in Williams-Beuren syndrome, a developmental disorder caused by deletion of multiple genes at 7q11.23.This gene encodes a member of the bromodomain protein family. The bromodomain is a structural motif characteristic of proteins involved in chromatin-dependent regulation of transcription. This gene is deleted in Williams-Beuren syndrome, a developmental disorder caused by deletion of multiple genes at 7q11.23.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BAZ1B
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.96 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene BAZ1B
Gene Accession No. Q9UIG0, Q9Z277
Gene Alias BAZ1B; bromodomain adjacent to zinc finger domain 1B; bromodomain adjacent to zinc finger domain protein 1B; bromodomain adjacent to zinc finger domain, 1B; bromodomain adjacent to zinc finger domain, 1B; Williams-Beuren syndrome chromosome region 9 homolog; C87820; hWALp2; transcription factor WSTF; tyrosine-protein kinase BAZ1B; tyrosine-protein kinase BAZ1B; LOW QUALITY PROTEIN: tyrosine-protein kinase BAZ1B; Tyrosine-protein kinase BAZ1B-like protein; WBSC10; WBSCR10; WBSCR9; williams syndrome transcription factor; Williams syndrome transcription factor homolog; williams-Beuren syndrome chromosomal region 10 protein; Williams-Beuren syndrome chromosomal region 9 protein; williams-Beuren syndrome chromosomal region 9 protein homolog; Williams-Beuren syndrome chromosome region 9 homolog; Wstf
Gene Symbols BAZ1B
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22385, 368002, 9031
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.