Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BAZ1B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103057
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103057 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103057 Supplier Invitrogen™ Supplier No. PA5103057
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total BAZ1B.

BAZ1B is a member of the bromodomain protein family. The bromodomain is a structural motif characteristic of proteins involved in chromatin-dependent regulation of transcription. This gene is deleted in Williams-Beuren syndrome, a developmental disorder caused by deletion of multiple genes at 7q11.23.This gene encodes a member of the bromodomain protein family. The bromodomain is a structural motif characteristic of proteins involved in chromatin-dependent regulation of transcription. This gene is deleted in Williams-Beuren syndrome, a developmental disorder caused by deletion of multiple genes at 7q11.23.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BAZ1B
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene BAZ1B
Gene Accession No. Q9UIG0
Gene Alias BAZ1B; bromodomain adjacent to zinc finger domain 1B; bromodomain adjacent to zinc finger domain protein 1B; bromodomain adjacent to zinc finger domain, 1B; bromodomain adjacent to zinc finger domain, 1B; Williams-Beuren syndrome chromosome region 9 homolog; C87820; hWALp2; transcription factor WSTF; tyrosine-protein kinase BAZ1B; tyrosine-protein kinase BAZ1B; LOW QUALITY PROTEIN: tyrosine-protein kinase BAZ1B; Tyrosine-protein kinase BAZ1B-like protein; WBSC10; WBSCR10; WBSCR9; williams syndrome transcription factor; Williams syndrome transcription factor homolog; williams-Beuren syndrome chromosomal region 10 protein; Williams-Beuren syndrome chromosomal region 9 protein; williams-Beuren syndrome chromosomal region 9 protein homolog; Williams-Beuren syndrome chromosome region 9 homolog; Wstf
Gene Symbols BAZ1B
Host Species Rabbit
Immunogen A synthesized peptide derived from human BAZ1B(Accession Q9UIG0), corresponding to amino acid residues P331-N381.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9031
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.