Learn More
Invitrogen™ BAZ1B Recombinant Rabbit Monoclonal Antibody (10Y9B6)

Description
Positive Samples: HeLa, PC-12, SH-SY5Y, SKOV3, Mouse brain Immunogen sequence: MAPLLGRKPF PLVKPLPGEE PLFTIPHTQE AFRTREEYEA RLERYSERIW TCKSTGSSQL THKEAWEEEQ EVAELLKEEF PAWYEKLVLE MVHHNTASLE.
Specifications
Specifications
| Antigen | BAZ1B |
| Applications | ELISA, Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | 10Y9B6 |
| Concentration | 0.25 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | BAZ1B |
| Gene Accession No. | Q9UIG0, Q9Z277 |
| Gene Alias | BAZ1B; bromodomain adjacent to zinc finger domain 1B; bromodomain adjacent to zinc finger domain protein 1B; bromodomain adjacent to zinc finger domain, 1B; bromodomain adjacent to zinc finger domain, 1B; Williams-Beuren syndrome chromosome region 9 homolog; C87820; hWALp2; transcription factor WSTF; tyrosine-protein kinase BAZ1B; tyrosine-protein kinase BAZ1B; LOW QUALITY PROTEIN: tyrosine-protein kinase BAZ1B; Tyrosine-protein kinase BAZ1B-like protein; WBSC10; WBSCR10; WBSCR9; williams syndrome transcription factor; Williams syndrome transcription factor homolog; williams-Beuren syndrome chromosomal region 10 protein; Williams-Beuren syndrome chromosomal region 9 protein; williams-Beuren syndrome chromosomal region 9 protein homolog; Williams-Beuren syndrome chromosome region 9 homolog; Wstf |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.