Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ BAZ2-ICR
GREENER_CHOICESDP

Catalog No. 526650 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-665-0 50 mg
52-661-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 52-665-0 Supplier Tocris Bioscience™ Supplier No. 5266/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective BAZ2 inhibitor

BAZ2-ICR is a selective BAZ2 bromodomain inhibitor (IC50 values are 130 and 180 nM for BAZ2A and BAZ2B respectively). Exhibits 15-fold selectivity for the BAZ2 bromodomain over the CERC2 bromodomain and >100-fold selectivity over a range of other bromodomains. Accelerates FRAP recovery in a BAZ2A FRAP assay.

Specifications

Chemical Name or Material 4-[4-(1-Methyl-1H-pyrazole-4-yl)-1-[2-(1-methyl-1H-pyrazol-4-yl)ethyl]-1H-imidazol-5-yl]benzonitrile
Quantity 50 mg
Target Bromodomain Inhibitors
Molecular Formula C20H19N7
Formula Weight 357.41
Purity >98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.