Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BAZ2A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP259158
Description
BAZ2A Polyclonal specifically detects BAZ2A in Human, Mouse samples. It is validated for Western Blot, Chromatin Immunoprecipitation.Specifications
BAZ2A | |
Polyclonal | |
Western Blot 1:1000, Chromatin Immunoprecipitation (ChIP) 5 uL/IP | |
bromodomain adjacent to zinc finger domain protein 2A, bromodomain adjacent to zinc finger domain, 2A, DKFZp781B109, FLJ45876, hWALp3, KIAA0314FLJ13768, Tip5, TIP5FLJ13780, Transcription termination factor I-interacting protein 5, TTF-I interacting peptide 5, TTF-I-interacting protein 5, WALp3 | |
Rabbit | |
Whole Antiserum | |
RUO | |
11176 | |
Store at -20C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, ChIP Assay | |
Unconjugated | |
Serum with 0.05% Sodium Azide | |
BAZ2A | |
The exact immunogen is propietary information. | |
100 μL | |
Primary | |
Human, Mouse | |
Antisera |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction