Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BAZ2A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120565
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120565 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120565 Supplier Invitrogen™ Supplier No. PA5120565
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: K-562, NIH/3T3. Immunogen sequence: QYPSANPGSN LKDPPLLSQF SGGQYPLNGI LGGSRQPSSP SHNTNLRAGS QEFWANGTQS PMGLNFDSQE LYDSFPDQNF EVMPNGPPSF FTSPQTSPML GSSIQTFAPS QEVGSGIHPD EAAEKEMTSV VAENGTGLVG SLELEEEQPE LKMCGYNGSV PSVESLHQEV SVLVPDPTVS CLDDPSHLPD QLEDTPILS.

TIP 5 is the large subunit of the nucleolar remodeling complex NoRC. NoRC causes the repression of ribosomal gene transcription. It was demonstrated that histone deacetylation is involved in this repression and that TIP5 is associated with the histone deacetylase HDAC1 and mediates the deacetylation of histones in the vicinity of the rDNA promoter. The interaction of TIP5 and HDAC1, which is necessary for transcriptional repression, is established by the C-terminal PHD finger and bromodomain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BAZ2A
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.16 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene BAZ2A
Gene Accession No. Q9UIF9
Gene Alias AA415431; BAZ2A; bromodomain adjacent to zinc finger domain 2A; bromodomain adjacent to zinc finger domain protein 2A; bromodomain adjacent to zinc finger domain, 2A; C030005G16Rik; C78388; hWALp3; KIAA0314; mKIAA0314; TIP5; Transcription termination factor I-interacting protein 5; TTF-I interacting peptide 5; TTF-I-interacting protein 5; Walp3
Gene Symbols BAZ2A
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 97-295 of human BAZ2A (NP_038477.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11176
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.