Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BAZ2A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5109445
Description
Immunogen sequence: QYPSANPGSN LKDPPLLSQF SGGQYPLNGI LGGSRQPSSP SHNTNLRAGS QEFWANGTQS PMGLNFDSQE LYDSFPDQNF EVMPNGPPSF FTSPQTSPML GSSIQTFAPS QEVGSGIHPD EAAEKEMTSV VAENGTGLVG SLELEEEQPE LKMCGYNGSV PSVESLHQEV SVLVPDPTVS CLDDPSHLPD QLEDTPILS.
TIP 5 is the large subunit of the nucleolar remodeling complex NoRC. NoRC causes the repression of ribosomal gene transcription. It was demonstrated that histone deacetylation is involved in this repression and that TIP5 is associated with the histone deacetylase HDAC1 and mediates the deacetylation of histones in the vicinity of the rDNA promoter. The interaction of TIP5 and HDAC1, which is necessary for transcriptional repression, is established by the C-terminal PHD finger and bromodomain.
Specifications
BAZ2A | |
Polyclonal | |
Unconjugated | |
BAZ2A | |
AA415431; BAZ2A; bromodomain adjacent to zinc finger domain 2A; bromodomain adjacent to zinc finger domain protein 2A; bromodomain adjacent to zinc finger domain, 2A; C030005G16Rik; C78388; hWALp3; KIAA0314; mKIAA0314; TIP5; Transcription termination factor I-interacting protein 5; TTF-I interacting peptide 5; TTF-I-interacting protein 5; Walp3 | |
Rabbit | |
Affinity chromatography | |
RUO | |
11176, 116848 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
2.48 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q91YE5, Q9UIF9 | |
BAZ2A | |
Recombinant fusion protein containing a sequence corresponding to amino acids 97-295 of human BAZ2A (TIP5) (NP_0384772). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction