Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BAZ2B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582699
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582699 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582699 Supplier Invitrogen™ Supplier No. PA582699
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NAGANGCTLS YQNSGKHSLG SVQSTATQSN VEKADSNNLF NTGSSGPGKF YSPLPNDQLL KTLTEKNRQW FSLLPRTPCD DTSLTHADMS TASLVTPQSQ PPSKS Highest antigen sequence indentity to the following orthologs: Rat - 75%, Mouse - 75%.

This gene belongs to the bromodomain gene family. Members of this gene family encode proteins that are integral components of chromatin remodeling complexes. The encoded protein showed strong preference for the activating H3K14Ac mark in a histone peptide screen, suggesting a potential role in transcriptional activation. This gene may be associated with susceptibility to sudden cardiac death (SCD). [provided by RefSeq, Aug 2016].
TRUSTED_SUSTAINABILITY

Specifications

Antigen BAZ2B
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BAZ2B
Gene Accession No. Q9UIF8
Gene Alias BAZ2B; bromodomain adjacent to zinc finger domain 2B; bromodomain adjacent to zinc finger domain protein 2B; bromodomain adjacent to zinc finger domain, 2B; DKFZP434H071; DKFZp762I0516; FLJ45644; hWALp4; KIAA1476; WALp4
Gene Symbols BAZ2B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human BAZ2B. Recombinant protein control fragment (Product #RP-91626).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29994
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.