Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BB515 Mouse Anti-Integrin α7 (ITGA7), BD Horizon™

Catalog No. BDB569837 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB569837 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB569837 Supplier BD Biosciences Supplier No. 569837
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Integrin α7 (ITGA7) Monoclonal antibody specifically detects Integrin α7 (ITGA7) in Mouse|Human samples. It is validated for Flow cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Integrin α7 (ITGA7)
Applications Flow Cytometry
Classification Monoclonal
Clone 3C12
Concentration 0.2 mg/mL
Conjugate Brilliant Blue 515 (BB515)
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias ITGA7; Integrin α7; integrin alpha-7; α7 Integrin
Host Species Mouse
Immunogen Mouse Myoblasts
Purification Method Affinity Chromatography
Quantity 0.1 mg
Regulatory Status RUO
Target Species Mouse, Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.