Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BB515 Mouse Anti-Mouse CD369 (CLEC7A), BD Horizon™

Catalog No. BDB569911 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB569911 25 μg
BDB569836 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB569911 Supplier BD Biosciences Supplier No. 569911
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD369 (CLEC7A) Monoclonal antibody specifically detects CD369 (CLEC7A) in Mouse samples. It is validated for Flow cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD369 (CLEC7A)
Applications Flow Cytometry
Classification Monoclonal
Clone 2F3
Concentration 0.2 mg/mL
Conjugate Brilliant Blue 515 (BB515)
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Clec7a; CD369; BGR; Beta-glucan receptor; Bgr; Dectin-1; Dectin1; dectin-1
Host Species Mouse
Immunogen Mouse Dectin-1 Recombinant Protein
Purification Method Affinity Chromatography
Quantity 25 μg
Regulatory Status RUO
Gene ID (Entrez) 56644
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.