Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD BBL™ Brucella Agar with 5% Sheep Blood, Hemin and Vitamin K1 Prepared Media
Click to view available options
Quantity:
100/Pk.
20/Pk.
Media Type:
Agar with 5% Sheep Blood, Hemin and Vitamin K1
Brucella Agar with 5% Sheep Blood, Hemin and Vitamin K1
Specifications
Specifications
Certifications/Compliance | ISO/DIS, 11014 |
Color | Red |
Content And Storage | 2°C to 8°C |
Form | Liquid |
Manufacturing Origin | United States |
Media Type | Brucella Agar with 5% Sheep Blood, Hemin and Vitamin K1 |
Formulation | Pancreatic digest of Casein 10g/L, Peptic digest of Animal Tissue 10g/L, Dextrose 1g/L, Yeast extract 2g/L, Sodium Chloride 5g/L, Sodium Bisulfite 0.1g/L, Agar 15g/L, Hemin 0.005g/L, Vitamin K1 0.01g/L, Sheep Blood, defibrinated 5% |
For Use With (Application) | Fastidious, obligately anaerobic organisms |
Packaging Type | Plate |
Quantity | 20/Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction