Learn More
Description
Certifications
AOAC, BAM, CCAM, COMPF, EPA, ISO, SMD, SMWW
Specifications
Specifications
| Certifications/Compliance | AOAC, BAM, CCAM, COMPF, EPA, ISO, SMD, SMWW |
| Content And Storage | Refrigerate |
| Form | Dehydrated |
| Media Type | Lauryl Sulfate Broth |
| Product Type | Nutrient Broth |
| Formulation | Pancreatic digest of Casein 20g/L, Lactose 5g/L, Dipotassium Phosphate 2.75g/L, Monopotassium Phosphate 2.75g/L, Sodium Chloride 5g/L, Sodium Lauryl Sulfate 0.1g/L |
| For Use With (Application) | Detection of Coliform bacteria |
| Packaging Type | Pail |
| Quantity | 11.3 kg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.