Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™/Difco™ Media Additives/Ingredients/Reagents: Phosphate Buffered Saline

Catalog No. p-4897966 Shop All BD Diagnostic Systems Products
Click to view available options
Quantity:
10 g
100 g
Packaging Type:
Bottle
Vial
This item is not returnable. View return policy
This item is not returnable. View return policy

Fluorescent Antibody reagents

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Content And Storage <30°C
Additive Phosphate Buffered Saline
Form Dehydrated
For Use With (Application) Fluorescent Antibody reagents
Packaging Type Bottle
Quantity 100 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.