Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Mycoflask™ Trypticase™ Soy Broth Prepared Media

Catalog No. p-4639709 Shop All BD Diagnostic Systems Products
Click to view available options
Quantity:
100 x 2 mL
100 x 5 mL
100 x 8 mL
100 x 10 mL
100 x 15 mL
Packaging Type:
Size A tubes with cap
Size K tubes with cap
This item is not returnable. View return policy
This item is not returnable. View return policy

Used for the cultivation of fastidious and nonfastidious microorganisms from a variety of clinical and nonclinical specimens

Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014, AOAC, BAM, BS12, CCAM, CLSI, CMPH2, COMPF, EP, EPA, ISO, JP, MCM9, SMD, USDA, USP
Color Beige
Content And Storage 15°C to 25°C
Form Liquid
Manufacturing Origin United States
Media Type Trypticase Soy Broth
Formulation Pancreatic digest of Casein 17g/L, Papaic digest of Soybean Meal 3g/L, Sodium Chloride 5g/L, Dipotassium Phosphate 2.5g/L, Dextrose 2.5g/L
For Use With (Application) For qualitative procedures for the cultivation of fastidious and non-fastidious microorganisms
Packaging Type Size K tubes with cap
Quantity 100 x 8 mL

For in vitro diagnostic use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.