Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD BBL™ Prepared Plated Media: Levine EMB Agar
Click to view available options
Quantity:
100/Pk.
Description
- Ready-to-use
- 100 x 15mm-style plates

Specifications
Specifications
Certifications/Compliance | ISO/DIS, 11014 |
Color | Light Beige |
Form | Liquid |
Manufacturing Origin | United States |
Media Type | Levine Eosin Methylene blue Agar |
Formulation | Pancreatic digest of Gelatin 10g/L, Lactose 10g/L, Dipotassium Phosphate 2g/L, Eosin Y 0.4g/L, Methylene blue 0.065g/L, Agar 15g/L |
For Use With (Application) | Gram negative enteric bacteria |
Packaging Type | Plate |
Quantity | 100/Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction