Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD BBL™ Prepared Plated Media: Sabouraud Dextrose Agar (Deep Fill)
Shop All BD Diagnostic Systems Products
Click to view available options
Quantity:
100/Pk.
20/Pk.
Description
- Ready-to-use
- 100 x 10mm and 100 x 15mm-style plates
- Deep-filled plates reduce effects of drying during prolonged incubation

Specifications
Specifications
Certifications/Compliance | USP, ISO/DIS, 11014 |
Color | Light Beige |
Form | Solid |
Manufacturing Origin | United States |
Media Type | Sabouraud Dextrose Agar (deep fill) |
Formulation | Pancreatic digest of Casein 5g/L, Peptic digest of Animal Tissue 5g/L, Dextrose 40g/L, Agar 15g/L |
For Use With (Application) | Qualitative procedures for cultivation of Dermatophytes, Candida albicans |
Packaging Type | Plate |
Quantity | 100/Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction