Learn More
BD BBL™ Sensi-Disc™ Antimicrobial Susceptibility Test Discs, Chloramphenicol29
Catalog No. p-4659972
Click to view available options
Concentration:
30 μg
Quantity:
1 Pack
10 Pack
This item is not returnable. View return policy
This item is not returnable. View return policy
Used for semi-quantitative in vitro testing by agar disc diffusion test procedure of certain fastidious bacterial pathogens
  • 6mm discs impregnated with accurately determined amounts of antibiotic or chemotherapeutic agents
  • Are clearly marked on both sides with letters and numbers designating agent and drug content
TRUSTED_SUSTAINABILITY
Specifications
Antimicrobial Agents Chloramphenicol29
Concentration 30 μg
Packaging Type Disc
Quantity 10 Pack
For in vitro diagnostic use
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.