Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BBOT, Scintillation Grade, Thermo Scientific™

Catalog No. AC401731000
Click to view available options
:
100g

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

For fluorescence/luminescence spectroscopy
TRUSTED_SUSTAINABILITY

Specifications

CAS 7128-64-5
Color Green to Yellow
Quantity 100g
Molecular Formula C26H26N2O2S
Infrared Spectrum Authentic
MDL Number MFCD00005774
Merck Index 15, 1012
Packaging Glass bottle
Synonym 2, 5-Bis(5'-tert-butyl-2-benzoxazol-2-yl)thiophene, 2, 5-Bis(5';-tert-butyl-2-benzoxazol-2-yl)thiophene
InChI Key AIXZBGVLNVRQSS-UHFFFAOYSA-N
SMILES CC(C)(C)C1=CC2=C(C=C1)OC(=N2)C3=CC=C(S3)C4=NC5=C(O4)C=CC(=C5)C(C)(C)C
IUPAC Name 5-tert-butyl-2-[5-(5-tert-butyl-1,3-benzoxazol-2-yl)thiophen-2-yl]-1,3-benzoxazole
Molecular Weight (g/mol) 430.566
PubChem CID 292429
Formula Weight 430.57
Percent Purity ≥99.0%
Chemical Name or Material BBOT
Show More Show Less
EINECSNumber 230-426-4
RTECSNumber DM4888332
TSCA TSCA
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.