Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BBOX1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA521477
Description
Recommended positive controls: Molt-4, mouse liver. Predicted reactivity: Mouse (86%), Rat (85%), Bovine (84%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
BBOX1 encodes gamma butyrobetaine hydroxylase which catalyzes the formation of L-carnitine from gamma-butyrobetaine, the last step in the L-carnitine biosynthetic pathway. Carnitine is essential for the transport of activated fatty acids across the mitochondrial membrane during mitochondrial beta-oxidation.
Specifications
BBOX1 | |
Polyclonal | |
Unconjugated | |
BBOX1 | |
AI874739; BBH; BBOX; Bbox1; butyrobetaine (gamma), 2-oxoglutarate dioxygenase (gamma-butyrobetaine hydroxylase) 1; butyrobetaine (gamma), 2-oxoglutarate dioxygenase 1 (gamma-butyrobetaine hydroxylase); gamma-BBH; Gamma-butyrobetaine dioxygenase; gamma-butyrobetaine hydroxylase; gamma-butyrobetaine hydroxylase 1; gamma-butyrobetaine,2-oxoglutarate dioxygenase; gamma-butyrobetaine,2-oxoglutarate dioxygenase 1; G-BBH | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
170442, 8424 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.76 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
O75936, Q924Y0 | |
BBOX1 | |
Recombinant protein encompassing a sequence within the center region of human BBOX1. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction