Learn More
Invitrogen™ BBOX1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA596433
Description
Immunogen sequence: MACTIQKAEA LDGAHLMQIL WYDEEESLYP AVWLRDNCPC SDCYLDSAKA RKLLVEALDV NIGIKGLIFD RKKVYITWPD EHYSEFQADW LKKRCFSKQA RAKLQRELFF PECQYWGSEL QLPTLDFEDV LRYDEHAYKW LSTLKKVGIV RLTGASDKPG EVSKLGKRMG FLYLTFYGHT; Positive Samples: U-251MG, MCF7, BT-474, Mouse liver, Mouse kidney, Mouse spinal cord; Cellular Location: Cytoplasm.
BBOX1 encodes gamma butyrobetaine hydroxylase which catalyzes the formation of L-carnitine from gamma-butyrobetaine, the last step in the L-carnitine biosynthetic pathway. Carnitine is essential for the transport of activated fatty acids across the mitochondrial membrane during mitochondrial beta-oxidation.
Specifications
BBOX1 | |
Polyclonal | |
Unconjugated | |
BBOX1 | |
AI874739; BBH; BBOX; Bbox1; butyrobetaine (gamma), 2-oxoglutarate dioxygenase (gamma-butyrobetaine hydroxylase) 1; butyrobetaine (gamma), 2-oxoglutarate dioxygenase 1 (gamma-butyrobetaine hydroxylase); gamma-BBH; Gamma-butyrobetaine dioxygenase; gamma-butyrobetaine hydroxylase; gamma-butyrobetaine hydroxylase 1; gamma-butyrobetaine,2-oxoglutarate dioxygenase; gamma-butyrobetaine,2-oxoglutarate dioxygenase 1; G-BBH | |
Rabbit | |
Affinity chromatography | |
RUO | |
170442, 64564, 8424 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
0.34 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O75936, Q924Y0, Q9QZU7 | |
BBOX1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-180 of human BBOX1 (NP_0039771). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.