Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ BBP33 Topcoated Polyimide Labels

Catalog No. 19107890 Shop All Brady Corporation Products
Click to view available options
Color:
White
Yellow
Width (English) Tape:
1.2 in.
1.7 in.
2.2 in.
3.2 in.

Mark wires and cables, particularly in aerospace defense and mass transit settings. Brady™ BBP33 Topcoated Polyimide Labels are self extinguishing labels with very high temperature ratings.

  • Can withstand very high temperatures
  • RoHS compliant
  • Used for wire and cable marking, particularly in aerospace defense and mass transit industries

Specifications

Certifications/Compliance RoHS
Description Chemical and humidity resistant
Adhesive Type Acrylic, Permanent
Color White
Finish Matte
Format Cartridge
Material Polyimide (B-472)
Product Type Topcoated Polyimide Label
For Use With (Equipment) BBP™33 Printer
Thickness (English) 0.0032 in.
Length (English) 85 ft.
Width (English) Tape 1.2 in.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.