Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BBS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113189
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113189 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113189 Supplier Invitrogen™ Supplier No. PA5113189
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BBS1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

Mutations in this gene have been observed in patients with the major form (type 1) of Bardet-Biedl syndrome. The encoded protein may play a role in eye, limb, cardiac and reproductive system development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BBS1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene BBS1
Gene Accession No. Q3V3N7, Q8NFJ9
Gene Alias AI451249; Bardet-Biedl syndrome 1; Bardet-Biedl syndrome 1 (human); Bardet-Biedl syndrome 1 homolog; Bardet-Biedl syndrome 1 protein; Bardet-Biedl syndrome 1 protein homolog; BBS1; BBS2L2; BBS2-like protein 2; D19Ertd609e
Gene Symbols BBS1
Host Species Rabbit
Immunogen Recombinant Human Bardet-Biedl syndrome 1 protein (217-417AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 52028, 582
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.