Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BBS1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 211181AP150UL

Description
Mutations in this gene have been observed in patients with the major form (type 1) of Bardet-Biedl syndrome. The encoded protein may play a role in eye, limb, cardiac and reproductive system development.Specifications
BBS1 | |
Polyclonal | |
Unconjugated | |
BBS1 | |
Bardet Biedl syndrome 1, BBS1, BBS2 like protein 2, BBS2L2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
52028, 582 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q3V3N7, Q8NFJ9 | |
Bbs1 | |
BBS1 Fusion Protein Ag15460 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction