Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BBS10 Antibody, Novus Biologicals™
SDP

Catalog No. p-7108632 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
20 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP168993 100 μL
NBP16899320 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP168993 Supplier Novus Biologicals Supplier No. NBP168993
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BBS10 Polyclonal specifically detects BBS10 in Human samples. It is validated for Western Blot.

Specifications

Antigen BBS10
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q8TAM1
Gene Alias Bardet-Biedl syndrome 10, Bardet-Biedl syndrome 10 protein, FLJ23560C12orf58chromosome 12 open reading frame 58
Gene Symbols BBS10
Host Species Rabbit
Immunogen Synthetic peptides corresponding to BBS10 (Bardet-Biedl syndrome 10) The peptide sequence was selected from the C terminal of BBS10. Peptide sequence SQTGLESVMGKYQLLTSVLQCLTKILTIDMVITVKRHPQKVHNQDSEDEL.
Molecular Weight of Antigen 81 kDa
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Vision
Primary or Secondary Primary
Gene ID (Entrez) 79738
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Rabbit
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.