Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BBS10 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563610
Description
Immunogen sequence: LEAYFCGRVG RNNHKFISQL MCDYFFKCMT CKSGIGVFEL VDDHFVELNV GVTGLPVSDS RIIAGLVLQK DFSVYRPADG DMRMVIVTET IQP Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 78%.
BBS10 is a member of the Bardet-Biedl syndrome (BBS) gene family. Bardet-Biedl syndrome is an autosomal recessive disorder characterized by progressive retinal degeneration, obesity, polydactyly, renal malformation and mental retardation. The proteins encoded by BBS gene family members are structurally diverse and the similar phenotypes exhibited by mutations in BBS gene family members is likely due to their shared roles in cilia formation and function. Many BBS proteins localize to the basal bodies, ciliary axonemes, and pericentriolar regions of cells. BBS proteins may also be involved in intracellular trafficking via microtubule-related transport. The protein encoded by this gene is likely not a ciliary protein but rather has distant sequence homology to type II chaperonins. As a molecular chaperone, this protein may affect the folding or stability of other ciliary or basal body proteins. Inhibition of this protein's expression impairs ciliogenesis in preadipocytes.
Specifications
BBS10 | |
Polyclonal | |
Unconjugated | |
BBS10 | |
1300007O09Rik; AI452285; Bardet-Biedl syndrome 10; Bardet-Biedl syndrome 10 (human); Bardet-Biedl syndrome 10 protein; Bardet-Biedl syndrome 10 protein homolog; Bbs10; C12orf58; RGD1560748; si:dkey-30c15.16 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79738 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8TAM1 | |
BBS10 | |
Recombinant protein corresponding to Human BBS10. Recombinant protein control fragment (Product #RP-97859). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction