Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BBS12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563989
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563989 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563989 Supplier Invitrogen™ Supplier No. PA563989
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EFEASTYIQH HLQNATDSGS PSSYILNEYS KLNSRIFNSD ISNKLEQIPR VYDVVTPKIE AWRRALDLVL LVLQTDSEII TGHGHTQINS QELTGFL Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 78%.

The BBS12 gene is a key contributor to Bardet-Biedl Syndrome (BBS), a rare genetic disorder characterized by primary cilia dysfunction, leading to diverse clinical manifestations such as retinal degeneration, obesity, polydactyly, renal abnormalities, and learning difficulties. BBS12 is located on chromosome 4q27 and encodes a vertebrate-specific chaperonin-like protein. Together with BBS6 and BBS10, BBS12 forms a part of a complex associated with the CCT/TRiC family of chaperonins, which are essential for the proper assembly of the BBSome complex. This complex plays a critical role in the trafficking of vesicles to cilia and is crucial for ciliary function and structure. Mutations in BBS12 are responsible for approximately 5% of BBS cases, underscoring its significant role in the pathophysiology of the syndrome. These mutations often lead to disruptions in ciliary functions, which are pivotal for the syndrome's manifestation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BBS12
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BBS12
Gene Accession No. Q6ZW61
Gene Alias Bardet-Biedl syndrome 12; Bardet-Biedl syndrome 12 (human); Bardet-Biedl syndrome 12 protein; Bardet-Biedl syndrome 12 protein homolog; Bbs12; C4orf24; DN-452A22.3; Gm1805; Gm407; Gm721
Gene Symbols BBS12
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human BBS12. Recombinant protein control fragment (Product #RP-103425).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 166379
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.