Learn More
Invitrogen™ BBS12 Polyclonal Antibody

Description
Recommended positive controls: HepG2. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | BBS12 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 |
| Gene | BBS12 |
| Gene Accession No. | Q6ZW61 |
| Gene Alias | Bardet-Biedl syndrome 12; Bardet-Biedl syndrome 12 (human); Bardet-Biedl syndrome 12 protein; Bardet-Biedl syndrome 12 protein homolog; Bbs12; C4orf24; DN-452A22.3; Gm1805; Gm407; Gm721 |
| Gene Symbols | BBS12 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.