Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BBS2 Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306335F
Description
BBS2 Polyclonal antibody specifically detects BBS2 in Human samples. It is validated for Western BlotSpecifications
BBS2 | |
Polyclonal | |
Western Blot | |
Bardet-Biedl syndrome 2, Bardet-Biedl syndrome 2 protein, BBS, MGC20703 | |
Produced in rabbits immunized with E. coli-derived Human BBS2 fragment. | |
0.1 mL | |
Vision | |
583 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
FITC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction