Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BBS4 Antibody (OTI2D5), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270241H
Description
BBS4 Monoclonal specifically detects BBS4 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
BBS4 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
BBS4 | |
Recombinant protein fragment corresponding to amino acids 266-519 of human BBS4 (NP_149017) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot | |
OTI2D5 | |
Western Blot | |
Bardet-Biedl syndrome 4, Bardet-Biedl syndrome 4 protein | |
Mouse | |
Affinity Purified | |
Stem Cell Markers, Vision | |
585.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction