Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BBS4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578617
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578617 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578617 Supplier Invitrogen™ Supplier No. PA578617
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: Neuro2A, C8D30, NIH-3T3, rat colon Predicted Reactivity: Human (100%), Pig (92%), Chimpanzee (98%), Bovine (91%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

BBS4 contains tetratricopeptide repeats (TPR), similar to O-linked N-acetyglucosamine transferase. Mutations in the gene encoding this protein have been observed in patients with Bardet-Biedl syndrome type 4. BBS4 may play a role in pigmentary retinopathy, obesity, polydactyly, renal malformation and mental retardation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BBS4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene BBS4
Gene Accession No. Q8C1Z7, Q96RK4
Gene Alias AW537059; AW742241; Bardet-Biedl syndrome 4; Bardet-Biedl syndrome 4 (human); Bardet-Biedl syndrome 4 homolog; Bardet-Biedl syndrome 4 protein; Bardet-Biedl syndrome 4 protein homolog; BBS4; D9Ertd464e; zgc:152964
Gene Symbols BBS4
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human BBS4. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 102774, 300754, 585
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.