Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BBS4 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA5142654
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:32,000.
BBS4 contains tetratricopeptide repeats (TPR), similar to O-linked N-acetyglucosamine transferase. Mutations in the gene encoding this protein have been observed in patients with Bardet-Biedl syndrome type 4. BBS4 may play a role in pigmentary retinopathy, obesity, polydactyly, renal malformation and mental retardation.
Specifications
BBS4 | |
Polyclonal | |
Unconjugated | |
BBS4 | |
AW537059; AW742241; Bardet-Biedl syndrome 4; Bardet-Biedl syndrome 4 (human); Bardet-Biedl syndrome 4 homolog; Bardet-Biedl syndrome 4 protein; Bardet-Biedl syndrome 4 protein homolog; BBS4; D9Ertd464e; zgc:152964 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
585 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q96RK4 | |
BBS4 | |
Peptide with sequence C-NEAAKLNQKDWEISH. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction